Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Hydrogen oxide

BDH23595.400 BDH23595.100
BDH23595.400CS 740.95 CAD
BDH23595.400 BDH23595.100
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Water

From the exclusive family of high-quality, competitively priced BDH Chemicals — high purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally friendly corrugated/pulp case inserts that are fully recyclable


These solvents are manufactured at a state of the art European ISO 9001/ISO 14001 solvent production facilities and feature an unparalleled combination of performance and price for even the most demanding liquid chromatography applications.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Filtered at 0.2 µm Passes Test
Coloration Max. 10 APHA
Evaporation Residue Max. 1 ppm
Spec. Conductivity (25°C) (at filling) Max. 1 µS/cm
Absorbance (200 nm) Max. 0.01
Absorbance (210 nm) Max. 0.01
Absorbance (220 nm) Max. 0.01
Absorbance (240 nm) Max. 0.01
Absorbance (254 nm) Max. 0.005
Absorbance (280 nm) Max. 0.005
Absorbance (300 nm) Max. 0.005
Fluorescence (as quinine) (254 nm) Max. 1 ppb
Fluorescence (as quinine) (365 nm) Max. 0.5 ppb
Gradient Grade (210 nm) Max. 5 mAU
Gradient Grade (254 nm) Max. 1 mAU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR