Methanol ≥99.8%, HiPerSolv CHROMANORM, gradient grade for HPLC, suitable for UPLC/UHPLC instruments, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filtered through a 0.2 µm filter, packaged under nitrogen.

BDH20864.400 BDH20864.100E
BDH20864.400CS 992.9 CAD
BDH20864.400 BDH20864.100E
Methanol ≥99.8%, HiPerSolv CHROMANORM, gradient grade for HPLC, suitable for UPLC/UHPLC instruments, VWR Chemicals BDH®
Methanol

From the exclusive family of high-quality, competitively priced BDH Chemicals – High Purity solvents for HPLC applications.


  • Designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Filtered down to 0.2 micron level and bottled under nitrogen
  • Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance
  • Feature environmentally friendly corrugated/pulp case inserts that are fully recyclable

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.8 %
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0002 meq/g
Evaporation residue Max. 5 ppm
Water Max. 0.03 %
Transmittance (220 nm) Min. 50 %
Transmittance (235 nm) Min. 80 %
Transmittance (260 nm) Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR