Ace-Thred Maxi Adapter for Trubore® Bearing, Ace Glass
Supplier: Ace Glass
8042-15 8042-17 8042-21 8042-35
89061-770EA 110.1 CAD
89061-770 89061-772 89061-776 89061-778
Ace-Thred Maxi Adapter for Trubore® Bearing, Ace Glass
Adapters Adapters, Spherical Ground Joints
This adapter is available in both "Midi" (#11 Ace-Thred) and "Maxi" (#15 Ace-Thred) formats.
It comes with a ground standard taper inner or spherical ball joint at bottom and an Ace-Thred joint at the top. The #11 Ace-Thred sizes will accept inner tubes with diameters of 9 to 10.5 mm, such as thermowells, gas dispersion tubes, vacuum takeoffs; the #15 Ace-Thred sizes will accept inner tubes with diameters of 12.5 mm to 14 mm such as electrodes, inlet and outlet tubes, etc. Suitable for most vacuum work. Available as complete item, with nylon bushing and FETFE o-ring, or as glassware only.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable