Powder Funnels, Globe Scientific
Supplier: Globe Scientific
600171 600167 600168 600169 600170 600166
10027-852CS 145.64 CAD
10027-852 10027-854 10027-856 10027-858 10027-860 10027-862
Powder Funnels, Globe Scientific
Funnels Powder Funnels
These heavy duty polypropylene (PP) funnels feature a large bore, which makes them ideal for use with powder, large particles and viscous liquids.
  • Heavy duty polypropylene (PP)
  • Autoclavable
  • Food safe
  • Air lock prevention
External ribbing is molded into the funnel to prevent air lock. They are angled at exactly 60° for rapid filtration.
Available in 6 different sizes, these funnels accommodate standard filter paper sizes.