Analytical Funnels, Globe Scientific
Supplier: Globe Scientific
600145 600150 600152 600153 600155 600156 600146 600147 600148
10027-832CS 95 CAD
10027-832 10027-834 10027-836 10027-838 10027-840 10027-842 10027-882 10027-884 10027-886
Analytical Funnels, Globe Scientific
Funnels Filtration Funnels Analytical Funnels
These heavy duty polypropylene (PP) analytical funnels are perfect for use in analytical chemistry applications
  • Heavy duty polypropylene (PP)
  • Autoclavable
  • Food safe
  • Air lock prevention
These analytical funnels have internal and external ribbing which helps to prevent air lock. They are angled at exactly 60° for rapid filtration.
Available in 9 different sizes, these analytical funnels accommodate standard filter paper sizes.
They feature long, narrow stems, which make them ideal for use with liquids.