Polypropylene Deep Well Plates, Biotix
Supplier: Biotix
63300107 63300108
89511-218CS 509.47 CAD
89511-218 89511-220 89511-222 89511-228 89511-234
Polypropylene Deep Well Plates, Biotix
Microplates Deep Well Microplates
Biotix deep well plates are made of non-treated medical-grade virgin PP.
  • Alphanumeric grid to help with sample identification
  • Notched corner as a reference point for plate orientation
  • Stackable for easy storage in freezers or incubators
  • Certified free of RNase, DNase, human DNA, Pyrogens and PCR Inhibitors
These plates are functional in a temperature range from –80 to 121°C with excellent chemical resistance. The plates conform to SBS standards to be used with automated workstations and can withstand up to 6,000 RCF in a microplate rotor.