SP Bel-Art Graduated Cylinders, Clear TPX®, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F28693-0000 F28696-0000 F28690-0000 F28692-0000 F28694-0000 F28695-0000 F28697-0000 F28691-0000 F28693-0000
13197-206EA 26.44 CAD
13197-206 13197-208 16126-580 16126-584 16126-586 24780-376 24780-378 30617-660 490014-142
SP Bel-Art Graduated Cylinders, Clear TPX®, Bel-Art Products, a part of SP
Cylinders Measuring Cylinders
Clear TPX® (PMP) plastic cylinders are non-wetting to prevent a concave meniscus, making the raised numeral, single scale graduations easy to read.
  • Superior chemical resistance
  • Shatter-proof
  • Safe for use to 175 ºC (350 ºF)
  • Steam autoclavable at 121 ºC (250 ºF)
  • Made of TPX® Brand polymethylpentene for easy viewing
They are shatter-proof with a wide octagonal base to prevent tipping and rolling.