Corning® LSE™ Orbital Shaker, Corning
Supplier: Corning
Corning®
6780-NP
89133-514CS 3302.23 CAD
89133-514
Corning® LSE™ Orbital Shaker, Corning
Shakers Microplate Shakers
The Corning LSE Orbital Shaker meets a variety of shaking and mixing applications
A powerful brushless motor and counter-balanced shaker mechanism provide smooth operation for years of trouble-free use. Different platforms (order separately) are available and are easily interchangeable. Digital control panel.
Certifications: CE Compliant, CSA Compliant.