NovaBlue Competent Cells, Cloning Strains, MilliporeSigma
Supplier: MilliporeSigma
69825-3 69825-4 71227-3 71227-4
CA80030-480EA 348.55 CAD
CA80030-480 CA80030-482 CA80601-638 CA80601-640
NovaBlue Competent Cells, Cloning Strains, MilliporeSigma
Competent Cells
NovaBlue Competent Cells are designed for convenient and reliable plasmid transformation
NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids), and recA and endA mutations, which result in high yields of excellent-quality plasmid DNA. Standard NovaBlue is ideal for routine cloning, while NovaBlue GigaSingles™ are most suitable for applications requiring higher transformation efficiencies.