Ettan™ IPGphor™ 3 Isoelectric Focusing System, Cytiva
Supplier: Cytiva
11002580 11002688 28933465 28933492 28938445 80649838 80649857
CA11028-332EA 2303.5 CAD
CA11028-332 CA11028-334 CA11028-336 CA11028-348 CA11028-350 CA11028-352 CA11028-398 CA11028-400
Ettan™ IPGphor™ 3 Isoelectric Focusing System, Cytiva
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
Ettan™ IPGphor™ 3 is a fully integrated isoelectric focusing (IEF) system optimized to deliver high throughput, speed, reproducibility and high protein-loading capacity.
  • Provides temperature control critical for reproducibility in 2-D DIGE experiments
  • Temperature control from 15 to 31 °C
  • Integrated high-voltage DC power supply delivering up to 10 kV makes the unit space-saving
  • Secure light protection during the run - Compatible with CyDye™-labelled proteins and other light-sensitive stains
  • Programming from the control panel or in Ettan™ IPGphor™ 3 Control
More Product Information