Sample and Storage Tubes, Globe Scientific
Supplier: Globe Scientific
6034 6050 6051 6052 6058 6059 6053 6054 6055 6030 6031 6032 6033 6035 6038 6039
89209-790CS 471.97 CAD
89209-790 10028-096 10028-098 10028-100 10028-102 10028-104 10028-180 10028-182 10028-184 76288-134 76288-136 76288-138 76288-140 76288-142 76288-144 76288-146
Sample and Storage Tubes, Globe Scientific
Tubes Test Tubes
Sample and storage tubes and caps have been designed for the storage and transportation of biological material.
  • Manufactured from non-toxic polypropylene
  • External threads provide a smooth and uniform inner surface
Manufactured from polypropylene for strength and clarity. The vial has external threads, which provide a smooth and uniform inner surface to reduce the risk of contamination. The screw caps are also produced from polypropylene, providing the same coefficient of expansion as the tubes. This, along with a polypropylene inner lip, ensures a leak-proof seal, even at changing temperatures. The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn.
The caps are available with or without a silicone O-ring, and are available in a variety of colors.
Ordering information: All screw caps are sold separately.