TLG® Separatory Funnels, Squibb, Pear-Shaped, Fully Autoclavable PTFE Stopcocks with Plastic Stopper, SATI International Scientific
Supplier: CHEM SCIENCE INC CA
CS-F0471330A CS-F0471660A CS-F0472125A CS-F0472250A CS-F0477500A CS-F0477000A
CA10823-560CS 1485.22 CAD
CA10823-560 CA10823-562 CA10823-564 CA10823-566 CA10823-568 CA10823-570
TLG® Separatory Funnels, Squibb, Pear-Shaped, Fully Autoclavable PTFE Stopcocks with Plastic Stopper, SATI International Scientific
Funnels Separating Funnels
TLG® Brand separatory funnels feature smooth tooled tips
Supplied with PTFE stopcocks to eliminate freezing, lubricant contamination, and to provide a precision fit within the barrel. The stopcock bores, body openings, and stems are carefully aligned for even liquid flow with fast cut-off. Made from heavy wall borosilicate 3.3 tubing with a low coefficient of expansion. Borosilicate tubing also provides uniform wall thickness.
Certifications: Designed from ASTM E-1096 Specifications.
Ordering information: All sizes are supplied with a polyethylene stopper.