Spherical Reaction Flask, 5 Neck, Ace Glass Incorporated
Supplier: Ace Glass
6470-20 6470-24 6470-26 6470-28
89054-520EA 3095.92 CAD
89054-520 89054-522 89054-524 89054-526
Spherical Reaction Flask, 5 Neck, Ace Glass Incorporated
Flasks Round Bottom Flasks
Rugged spherical reaction flask with 65/40 spherical joint center neck, two 65/40 spherical joint side necks and two 29/42 standard taper side joints
Made of borosilicate glass