TLG® Round Bottom Boiling Flasks, Heavy Wall, Single Neck, SATI Glassware
Supplier: CHEM SCIENCE INC CA
CS-F0301450 CS-F0301950 CS-F0302450 CS-F0301100 CS-F0309100 CS-F0304100 CS-F0301250 CS-F0309250 CS-F0304250 CS-F0301500 CS-F0309500 CS-F0304500 CS-F0304000 CS-F030242L CS-F0301405 CS-F0301410 CS-F0301910 CS-F0301425 CS-F0301925 CS-F0302425
CA10823-118CS 1000 CAD
CA10823-118 CA10823-120 CA10823-122 CA10823-124 CA10823-126 CA10823-200 CA10823-202 CA10823-204 CA10823-206 CA10823-208 CA10823-210 CA10823-212 CA10823-214 CA10823-216 CA10823-606 CA10823-608 CA10823-610 CA10823-612 CA10823-614 CA10823-616
TLG® Round Bottom Boiling Flasks, Heavy Wall, Single Neck, SATI Glassware
Flasks Round Bottom Flasks
TLG® Brand boiling flasks are designed from ASTM Specification E1403 requirements
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness. Made from Borosilicate 3.3 glass.
Certifications: ASTM Specification E1403.