TLG® Heavy-Duty Low Form Beakers, Graduated, SATI glassware
Supplier: CHEM SCIENCE INC CA
229.203.16 229.203.07 229.203.08 229.203.10 229.203.11 229.203.14
CA10822-970CS 1225.04 CAD
CA10822-970 CA10823-194 CA10823-196 CA10823-700 CA10823-702 CA10823-704 CA10823-706
TLG® Heavy-Duty Low Form Beakers, Graduated, SATI glassware
Beakers
TLG® Brand heavy duty beakers made with thick uniform walls are intended to prolong life expectancy under harsher conditions than the normal usage (i.e mechanical shock).
  • 250 to 4000ml sizes are marked
  • Evenly tooled top rim
  • Spout is designed for optimum performance
All sizes have white, permanent graduations, and large marking squares.
Certifications: Designed from ASTM Specification E960 requirements.
Ordering information: Not recommended for use on hot plates where significant thermal gradients exist due to lack of agitation of contents orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable