TLG® Griffin Low Form Beakers with Double-Capacity Scale, SATI Glassware
Supplier: SATI International Scientific
229.202.06 229.202.04 229.202.05 229.202.10 229.202.11 229.202.03 229.202.02 229.202.08 229.202.07 229.202.02A
CA10823-544CS 1004.01 CAD
CA10823-544 CA10823-742 CA10823-546 CA10823-744 CA10823-740 CA10823-542 CA10823-708 CA10823-548 CA10823-746 CA10823-748
TLG® Griffin Low Form Beakers with Double-Capacity Scale, SATI Glassware
Beakers
TLG® Brand Beakers meet ASTM E-960 Specifications for classification, design, capacity & dimensions, and markings
  • Easy-pour spouts
  • White, permanent graduations
  • Large marking squares
  • Made from high-quality borosilicate glass with a low coefficient of expansion
Manufactured with uniform wall thickness resulting in the optimum balance between thermal shock resistance and mechanical strength. 20mL through 4000mL beakers have double-graduated metric scales. Accuracy: ±5% of full capacity.
Certifications: ASTM E-960.