SP Bel-Art Buchner Funnels, Single Piece, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F14608-0000 F14609-0000 F14610-0000 F14611-0000
70690-252CS 617.39 CAD
70690-252 70690-254 70690-256 70690-258
SP Bel-Art Buchner Funnels, Single Piece, Bel-Art Products, a part of SP
Funnels Filtration Funnels Büchner Funnels
One-piece HDPE plastic funnels with sealed-in, polyethylene filter plate of coarse porosity (90-130 microns) offer good retention and fast flow under vacuum.
  • One-piece design
  • HDPE funnel construction
  • Polyethylene filter plate
  • Good retention and fast flow
Useable with or without filter paper. Use below 52ºC (125ºF). Use with Vac-Ring Filter Seal (H38959-0000) for quick filter setup.
Order Now
SPECIFICATIONS
Environmentally Preferable