SP Bel-Art Standard Stem Funnels, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F14676-0055 F14676-0065 F14676-0075 F14676-0100
47750-830PK 187.66 CAD
47750-830 30617-614 30617-616 30617-618
SP Bel-Art Standard Stem Funnels, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Polypropylene plastic funnels feature an exclusive filter-step internal design for speedier filtrations.
  • Polypropylene plastic construction
  • External ribs
  • Autoclavable
  • Filter-step internal design
External ribs prevent airlock. The parallel stem measures 8 mm O.D. Steam autoclavable at 121ºC (250ºF).