Glass Precision Ground Button Type Stirrer Shaft, 19 mm, Ace Glass
Supplier: Ace Glass
8077-23 8077-25 8077-27
89061-986EA 401.85 CAD
89061-986 89061-988 89062-008
Glass Precision Ground Button Type Stirrer Shaft, 19 mm, Ace Glass
Stirrers Stirring Shafts
The 19 mm O.D
button type with a hollow shaft is for use with all 19 mm I.D. ACE Trubore® bearings. Also compatible with a mechanical seal and PTFE bearings. Fits certain ACE chucks. Ground surface approximately 380 mm long.