BugBuster® HT Protein Extraction Reagent, MilliporeSigma
Supplier: MilliporeSigma
70922-3 70922-4 70922-5
CA80500-206EA 423.55 CAD
CA80500-206 CA80500-208 CA80509-638
BugBuster® HT Protein Extraction Reagent, MilliporeSigma
Protein Extraction Protein Extraction Reagents
BugBuster® HT Protein Extraction Reagent is formulated for the gentle disruption of the cell wall of E
BugBuster® HT is a ready-to-use, premixed formulation of BugBuster and Benzonase® Nuclease ideally suited to high-throughput applications.
coli, resulting in the liberation of soluble protein. It provides a simple, rapid, low cost alternative to mechanical methods such as French Press or sonication for releasing expressed target proteins in preparation for functional studies or purification.