Plate Count Agar, HiMedia
Supplier: HIMEDIA LABORATORIES PVT. CA
M091-2.5KG
M091A-500G
M091S-500G
M091-5KG
M091-500G
M091A-2.5KG
M091A-5KG
M091S-2.5KG
M091S-5KG
CA89135-058EA
3498.47
CAD
CA89135-058
CA95020-240
CA95020-242
CA95026-808
CA95020-238
CA89222-522
CA89222-524
CA89222-526
CA89222-528
Plate Count Agar, HiMedia
Media for Microbiology
Dehydrated Media for Microbiology
Plate Count Agar is recommended for the determination of plate counts of microorganisms in food, water and waste water.
Cream to yellow, homogeneous free flowing powder