BugBuster® Protein Extraction Reagent, MilliporeSigma
Supplier: MilliporeSigma
70584-3 70584-4
CA80031-732EA 259.04 CAD
CA80031-732 CA80031-734
BugBuster® Protein Extraction Reagent, MilliporeSigma
Protein Extraction Protein Extraction Reagents
BugBuster® Protein Extraction Reagent is formulated for the gentle disruption of the cell wall of E.coli, resulting in the liberation of soluble protein.
It provides a simple, rapid, low cost alternative to mechanical methods such as French Press or sonication for releasing expressed target proteins in preparation for functional studies or purification.