SP Bel-Art ProCulture Cryogenic Vial Storage Boxes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18849-0001 F18849-0002 F18849-0000
47751-740PK 110.96 CAD
47751-740 47751-742 47751-738
SP Bel-Art ProCulture Cryogenic Vial Storage Boxes, Bel-Art Products, a part of SP
Boxes Cryoboxes
Durable polycarbonate boxes are ideal for the compact storage of cell stocks in 1.2, 2.0, or 5.0 mL cryogenic vials.
  • Can be used in temperatures from –196 ºC to +121 ºC
  • Clear lids can be written on with permanent markers and have a printed grid for easy inventory
  • Keyed corner aligns the lid with identifying numbers on the rack
  • Useful as mailers for vials
Box 47751-740 can be used with Cryo Tower Storage Systems (74950-132 and 74950-134) to save valuable bench space.