Tricorn Empty Chromatography Columns
Supplier: Cytiva
28-4064-11 28-4064-08 28-4064-13 28-4064-19 28-4064-12 28-4064-10 28-4064-14 28-4064-16 28-4064-17 28-4064-18 28-4064-09 28-4064-15
CA10768-776EA 1894.14 CAD
CA10768-776 CA10768-778 CA10768-780 CA10768-782 CA10768-784 CA10768-786 CA10768-788 CA10768-790 CA10768-792 CA10768-794 CA10768-796 CA10768-798
Tricorn Empty Chromatography Columns
Chromatography Columns
Tricorn empty columns are for high resolution small-scale chromatography at pressures up to 50 bar (Tricorn 10) or 100 (Tricorn 5).
  • Versatile – designed for high resolution liquid chromatography on a small and medium scale (up to 47.6 mL bed volume)
  • High pressure stability – up to 50 bar (Tricorn 10) and 100 bar (Tricorn 5), for reproducible use and high flow rates
  • Broadly compatible – functions well with all Cytiva chromatography resins and most other commercial lab resins
  • Simple operation – integrated, snap-on design facilitates assembly, operation, and
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable