Support Stand for Big Columns, Ace Glass Incorporated
Supplier: Ace Glass
11056-20 11056-35
89075-604EA 4408.85 CAD
89075-604 89075-606
Support Stand for Big Columns, Ace Glass Incorporated
Retort Stands
Support stand for larger columns with #50 or #80 Ace-Threads
Three threaded holes are present in base legs to accommodate rods.
Delivery information: Supplied with stainless steel U shaped base, one 5/8" x 5' rod, two 5/8" x 29 rods, three power hold clamps, three 11079-08 chain clamps, a modified 7837 aluminum support stand (18" high), and one 11750 cork ring.