Glass Precision Ground Knob Type Stirrer Shaft, 19 mm, Ace Glass
Supplier: Ace Glass
8078-05 8078-10
89062-012EA 629.07 CAD
89062-012 89062-014
Glass Precision Ground Knob Type Stirrer Shaft, 19 mm, Ace Glass
Stirrers Stirring Shafts
Precision ground glass stirring shafts, 19 mm O.D., hollow, heavy duty
Knob type shafts accept agitators and can be used in all 19 mm I.D. and Trubore® bearings. Compatible with ACE mechanical seal and PTFE debris-free bearings. Join to stirrer motor using series chucks. Heavy duty design intended for use with large flasks up to 72 liters.
Accessories information: PTFE Gaskets are designed to fit a variety of stirring shafts, such as 89061-938, 89061-946, 89084-572, 89061-962, or 89062-014. Gaskets are also useful as a dust cover, or as a replacement for the gasket supplied withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable