Heat Exchanger Cooling/Heating Coil Accessories, Ace Glass
Supplier: Ace Glass
12067-239 12067-240 12067-244 12067-248 12067-261 12067-263 12067-265 12067-268 12067-279 12067-280 12067-39 12067-40 12067-44 12067-48 12067-52 12067-58 12067-61 12067-63 12067-65 12067-68 12067-71 12067-79 12067-80 12067-84
89076-234EA 1674.53 CAD
89076-234 89076-236 89076-238 89076-240 89076-244 89076-246 89076-248 89076-250 89076-252 89076-254 89076-256 89076-258 89076-262 89076-280 89076-284 89076-290 89076-292 89076-294 89076-296 89076-298 89076-300 89076-302 89076-304 89076-306
Heat Exchanger Cooling/Heating Coil Accessories, Ace Glass
Stirrers Stirring Tools
These are PTFE coated copper, Hastelloy C-276 or stainless steel coils for use with pilot plant reactors with either cylindrical or spherical flasks that contains 200 mm or 300 mm size flanges.
Coils are ideal for steam heating or water cooling of reactor contents. Available in 1/4" or 1/2" outer diameter sizes.