Glas-Col® Low Profile Aluminum Housed Heating Mantle, Ace Glass
Supplier: Ace Glass
12050-34 12050-36 12050-38 12050-41 12050-43 12050-45
89076-006EA 3799.57 CAD
89076-006 89076-008 89076-010 89076-012 89076-014 89076-016
Glas-Col® Low Profile Aluminum Housed Heating Mantle, Ace Glass
Heaters Heating Mantles
This mantle is used with reaction assemblies and is CSA certified
Easily fits into ACE 4-post reactor stands. Temperature range is Ambient 10°C to 450°C. Must be operated through a temperature controller. With detachable 4' cord and locking connector. Available with or without bottom opening.