Multi-Paddle Agitator, 10 mm, Ace Glass Incorporated
Supplier: Ace Glass
8094-23 8094-50 8094-52 8094-27
89062-178EA 686.38 CAD
89062-178 89062-182 89062-184 89062-188
Multi-Paddle Agitator, 10 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
For use with a stainless steel shaft or a borosilicate straight shaft
Center hole has a 10 mm inner diameter. and one or more can be positioned on shaft at various locations using set screws to secure. With stainless shaft use stainless set screw; with glass shaft use Kel-F-tipped set screw. Both screws supplied with agitator.