Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
Supplier: Ace Glass
8080-14 8080-18 8080-24 8080-25 8080-29 8080-12 8080-30 8080-16
89062-020EA 1417.82 CAD
89062-020 89062-022 89062-026 89062-028 89062-030 89097-174 89097-176 76107-028
Heavy Duty Polished Glass Stirrer Shaft, 28 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Heavy duty, 28 mm O.D
For Agitators and Turbines Reference the Following Numbers: 89062-152, 89062-174, 89062-176, 71001-576, 71001-578.
glass stirring shaft for use with ACE reactors. Shaft has pre-drilled holes at top and bottom for positive attachment to motor coupling and agitator, respectively. Recommended for use with ACE PTFE Trubore® and mechanical seal bearings. Use with agitators for 28 mm rods. Fits Ace 89054-598 swivel connector and 89054-602 chuck.
Order Now
SPECIFICATIONS
Environmentally Preferable