Covered Stainless Steel Stirrer Shaft, PTFE, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
8079-05 8079-10 8079-03
89062-016EA 1708.11 CAD
89062-016 89062-018 89084-574
Covered Stainless Steel Stirrer Shaft, PTFE, 19 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
Precision PTFE covered stainless steel stirring shafts, 19 mm O.D
Drilled type shafts accept agitators and can be used in 19 mm Trubore® bearings. Join to stirrer motor using certain ACE series chucks.