KOD Hot Start Master Mix, MilliporeSigma
Supplier: MilliporeSigma
KOD Hot Start Master Mix is a ready-to-use 2X mixture optimized for convenient high-fidelity PCR
The mix contains KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4. The Master Mix simplifies PCR set-up, offering time savings, consistency, and minimal risk of contamination. The mix is ideal for use in high-throughput applications. Simply add KOD Hot Start Master Mix to an equal volume of sample containing DNA template and primers.