Manganese(III) acetate dihydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
47050-04
47050-09
CAAA47050-04EA
235.42
CAD
CAAA47050-04
CAAA47050-09
Manganese(III) acetate dihydrate 97%
Manganese (III) acetate dihydrate
Manganese(III) acetate dihydrate, 97%
Formula:
(H₃CCOO)₂Mn·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150022 CAS Number: 19513-05-4 EINECS: 213-602-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...