Scandium(III) bis(trifluoromethylsulfonyl)imide, solid
Supplier: Thermo Scientific Chemicals
|
Danger
|
46948-03
46948-MD
CAAA46948-03EA
401.32
CAD
CAAA46948-03
CAAA46948-MD
Scandium(III) bis(trifluoromethylsulfonyl)imide, solid
Scandium(III) bis(trifluoromethylsulfonyl)imide
Scandium(III) bis(trifluoromethylsulfonyl)imide
CAS No.: 176726-07-1
Molecular Formula: C6F18S6O12N3Sc
Formula Weight: 885.37
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
Melting Point: 199-204°
UN No.: 1759
MDL No.: MFCD03427000
Formula:
C₆F₁₈N₃O₁₂S₆Sc Melting Pt: 199…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427000 CAS Number: 176726-07-1 UN: 1759 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...