Silver bis(trifluoromethanesulfonyl)imide
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Silver Triflimide, Silver bis(trifluoromethane sulphonimide), Silver bis(trifluoromethane sulfonimide)
46784-03
46784-06
46784-MD
CAAA46784-03EA
254.38
CAD
CAAA46784-03
CAAA46784-06
CAAA46784-MD
Silver bis(trifluoromethanesulfonyl)imide
Silver bis(trifluoromethanesulfonyl)imide
Silver bis(trifluoromethylsulfonyl)imide
CAS No.: 189114-61-2
Molecular Formula: C2AgF6NO4S2
Formula Weight: 388.00
Physical Form: Solid
Sensitivity: Moisture and Light Sensitive
Appearance: White to orange
Melting Point: 249°
UN No.: 1759
MDL No.: MFCD08457644
Formula:
C₂AgF₆NO₄S₂ MW: 388.00 g/mol Melting Pt: 249 °C Storage Temperature: Ambient |
MDL Number:
MFCD08457644 CAS Number: 189114-61-2 UN: 1759 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...