Calcium bis(trifluoromethylsulfonyl)imide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Calcium(II) Bis(trifluoromethanesulphonyl)imide, Calcium(II) Bis(trifluoromethanesulfonyl)imide, Calcium bis(trifluoromethanesulphonimide), Bis(trifluoromethanesulfonyl)imide calcium(II) salt, Bis(trifluoromethanesulphonyl)imide calcium(II) salt, Calcium(II) triflimide, Calcium bis(trifluoromethanesulfonimide), Calcium triflimide

46533-03 46533-06 46533-MD
CAAA46533-03EA 218.04 CAD
CAAA46533-03 CAAA46533-06 CAAA46533-MD
Calcium bis(trifluoromethylsulfonyl)imide
Calcium bis(trifluoromethylsulfonyl)imide

Calcium bis(trifluoromethylsulfonyl)imide


CAS No.: 165324-09-4 Molecular Formula: C4CaF12N2O8S4 Formula Weight: 600.35 Physical Form: Solid Sensitivity: Moisture Sensitive Melting Point: 300° UN No.: 1759 MDL No.: MFCD14156019

Formula: C₄CaF₁₂N₂O₈S₄
MW: 600.35 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD14156019
CAS Number: 165324-09-4
UN: 1759
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR