Ruthenium 4% on alumina, rings
Supplier: Thermo Scientific Chemicals
44593-36
44593-22
CAAAA44593-36EA
3431.76
CAD
CAAAA44593-36
CAAA44593-22
Ruthenium 4% on alumina, rings
Ruthenium
Ruthenium, 4% on 6.35mm (0.25in) alumina rings
CAS No.: 7440-18-8
Appearance: Greyish-white
MDL No.: MFCD00011207
Formula:
Ru MW: 101.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...