Tellurium dioxide ≥99.995% (metals basis), powder

Supplier: Thermo Scientific Chemicals
Warning

11821-09 11821-18 11821-30
CAAA11821-09EA 124.66 CAD
CAAA11821-09 CAAA11821-18 CAAA11821-30
Tellurium dioxide ≥99.995% (metals basis), powder
Tellurium dioxide

Tellurium(IV) oxide, 99.995% (metals basis)

Formula: O₂Te
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 733 °C
Density: 5.8
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,9123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR