Bis(p-tolyl)phosphine oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61504-03
H61504-06
CAAAH61504-03EA
79
CAD
CAAAH61504-03
CAAAH61504-06
Bis(p-tolyl)phosphine oxide 98%
Bis(p-tolyl)phosphine oxide
Bis(p-tolyl)phosphine oxide, 98%
Purity: 98%
CAS No.: 2409-61-2
Molecular Formula: C14H15OP
Formula Weight: 230.29
Physical Form: Solid
Appearance: White
Beilstein Reference: 2804632
MDL No.: MFCD01445489
Formula:
C₁₄H₁₅OP Storage Temperature: Ambient |
MDL Number:
MFCD01445489 CAS Number: 2409-61-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...