4-Fluoro-1,3-dioxolan-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Fluoroethylene Carbonate, 4-Fluoro-2-oxo-1,3-dioxolane

H61502-14 H61502-22
CAAAH61502-14EA 278.08 CAD
CAAAH61502-14 CAAAH61502-22
4-Fluoro-1,3-dioxolan-2-one 98%
4-Fluoro-1,3-dioxolan-2-one

4-Fluoro-1,3-dioxolan-2-one, 98%


Purity: 98% CAS No.: 114435-02-8 Molecular Formula: C3H3FO3 Formula Weight: 106.05 Storage Temperature: 2°C to 8°C Flash Point: 102°(216°F) Physical Form: Liquid Appearance: Colorless Melting Point: 18-23° Boiling Point: 212° MDL No.: MFCD06247543

Formula: C₃H₃FO₃
MW: 106.05 g/mol
Boiling Pt: 212 °C
Melting Pt: 18…23 °C
Flash Pt: >102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD06247543
CAS Number: 114435-02-8
EINECS: 483-360-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR