2,6-Dibromopyrazine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61468-03
H61468-06
CAAAH61468-03EA
126.56
CAD
CAAAH61468-03
CAAAH61468-06
2,6-Dibromopyrazine 95%
2,6-Dibromopyrazine
2,6-Dibromopyrazine, 95%
Purity: 95%
CAS No.: 23229-25-6
Molecular Formula: C4H2Br2N2
Formula Weight: 237.88
Storage Temperature: -30°C to -10°C
Physical Form: Crystalline solid
Appearance: Off-white to pale brown
MDL No.: MFCD08061601
Formula:
C₄H₂Br₂N₂ MW: 237.882 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08061601 CAS Number: 23229-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...