1-Bromo-5-fluoro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

H61466-06 H61466-14
CAAAH61466-06EA 73.79 CAD
CAAAH61466-06 CAAAH61466-14 CAAAH61466-22
1-Bromo-5-fluoro-2-nitrobenzene 98%
1-Bromo-5-fluoro-2-nitrobenzene

2-Bromo-4-fluoro-1-nitrobenzene, 98%

Formula: C₆H₃BrFNO₂
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00792441
CAS Number: 700-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR