4-Nitro-α,α,α-trifluoro-o-toluic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H61452-03 H61452-06
CAAAH61452-03EA 89.9 CAD
CAAAH61452-03 CAAAH61452-06
4-Nitro-α,α,α-trifluoro-o-toluic acid 97%
4-Nitro-α,α,α-trifluoro-o-toluic acid

4-Nitro-2-(trifluoromethyl)benzoic acid, 97%

Formula: C₈H₄F₃NO₄
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD04039246
CAS Number: 320-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR