4-Amino-5-bromo-2-chloropyrimidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61429-06
H61429-14
CAAAH61429-03EA
179.38
CAD
CAAAH61429-03
CAAAH61429-06
CAAAH61429-14
4-Amino-5-bromo-2-chloropyrimidine 95%
4-Amino-5-bromo-2-chloropyrimidine
4-Amino-5-bromo-2-chloropyrimidine, 95%
Purity: 95%
CAS No.: 205672-25-9
Molecular Formula: C4H3BrClN3
Formula Weight: 208.44
Physical Form: Solid
MDL No.: MFCD07782040
Formula:
C₄H₃BrClN₃ MW: 208.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782040 CAS Number: 205672-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...