2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61407-03
H61407-06
H61407-MD
CAAAH61407-03EA
137.46
CAD
CAAAH61407-03
CAAAH61407-06
CAAAH61407-MD
2,3-Dibromo-5-(Trifluoromethyl)Pyridine 95%
2,3-Dibromo-5-(Trifluoromethyl)Pyridine
2,3-Dibromo-5-(trifluoromethyl)pyridine, 95%
Purity: 95%
CAS No.: 79623-38-4
Molecular Formula: C6H2Br2F3N
Formula Weight: 304.89
Physical Form: Solid
MDL No.: MFCD10687233
Formula:
C₆H₂Br₂F₃N MW: 304.892 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10687233 CAS Number: 79623-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...