Pyrimidine-5-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H61406-03 H61406-06 H61406-MD
CAAAH61406-03EA 211.72 CAD
CAAAH61406-03 CAAAH61406-06 CAAAH61406-MD
Pyrimidine-5-carboxylic acid 95%
Pyrimidine-5-carboxylic acid

Pyrimidine-5-carboxylic acid, 95%


Purity: 95% CAS No.: 4595-61-3 Molecular Formula: C5H4N2O2 Formula Weight: 124.10 Physical Form: Solid Melting Point: 259-261° MDL No.: MFCD00856162

Formula: C5H4N2O2
MW: 124.10 g/mol
Melting Pt: 259…261 °C
Storage Temperature: Ambient
MDL Number: MFCD00856162
CAS Number: 4595-61-3
EINECS: 224-994-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR