4-Methoxy-6-methyl-2H-pyran-2-one 97%

Supplier: Thermo Scientific Chemicals

H61395-03 H61395-MD
CAAAH61395-03EA 159.58 CAD
CAAAH61395-03 CAAAH61395-MD
4-Methoxy-6-methyl-2H-pyran-2-one 97%
4-Methoxy-6-methyl-2H-pyran-2-one

4-Methoxy-6-methyl-2H-pyran-2-one, 97%


Purity: 97% CAS No.: 672-89-9 Molecular Formula: C7H8O3 Formula Weight: 140.14 Physical Form: Solid Melting Point: 83-86° MDL No.: MFCD00006640

Formula: C₇H₈O₃
MW: 140.138 g/mol
Melting Pt: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00006640
CAS Number: 672-89-9
EINECS: 211-600-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR