Methyl-3-(2-bromophenyl)propionate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(2-Bromophenyl)propionic Acid Methyl Ester, Methyl 3-(2-bromophenyl)propanoate
H61342-06
H61342-14
CAAAH61342-06EA
156.1
CAD
CAAAH61342-06
CAAAH61342-14
Methyl-3-(2-bromophenyl)propionate 98%
Methyl-3-(2-bromophenyl)propionate
Methyl 3-(2-bromophenyl)propionate, 98%
Purity: 98%
CAS No.: 66191-86-4
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Physical Form: Liquid
Appearance: Colorless to pale yellow
MDL No.: MFCD09953149
Formula:
C₁₀H₁₁BrO₂ MW: 243.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953149 CAS Number: 66191-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...