Diisopropylamine dichloroacetate 95%

Supplier: Thermo Scientific Chemicals
Warning

H61341-03 H61341-MD
CAAAH61341-03EA 181.7 CAD
CAAAH61341-03 CAAAH61341-06 CAAAH61341-MD
Diisopropylamine dichloroacetate 95%
Diisopropylamine dichloroacetate

Diisopropylamine dichloroacetate, 95%


Purity: 95% CAS No.: 660-27-5 Molecular Formula: C6H15N.C2H2Cl2O2 Formula Weight: 230.13 Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00868288

Formula: C₈H₁₇Cl₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD00868288
CAS Number: 660-27-5
EINECS: 211-538-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR