1,3-Dichloro-2-fluoro-5-nitrobenzene 95%

Supplier: Thermo Scientific Chemicals
Warning

H61321-06 H61321-14
CAAAH61321-06EA 69.2 CAD
CAAAH61321-06 CAAAH61321-14 CAAAH61321-22
1,3-Dichloro-2-fluoro-5-nitrobenzene 95%
1,3-Dichloro-2-fluoro-5-nitrobenzene

3,5-Dichloro-4-fluoro-1-nitrobenzene, 95%

Formula: C₆H₂Cl₂FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00129963
CAS Number: 3107-19-5
EINECS: 221-466-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR